EMAIL THIS PAGE TO A FRIEND
Chemosphere
Occurrence of phthalate diesters (phthalates), p-hydroxybenzoic acid esters (parabens), bisphenol A diglycidyl ether (BADGE) and their derivatives in indoor dust from Vietnam: Implications for exposure.
PMID 26498104
Abstract
Phthalate diesters (phthalates), esters of p-hydroxybenzoic acid (parabens), and bisphenol A diglycidyl ether (BADGE) are used in personal care products, food packages, household products, or pharmaceuticals. These compounds possess endocrine-disrupting potentials and have been reported to occur in the environment. Nevertheless, no previous studies have reported the occurrence of these compounds in indoor dust from Vietnam. In this study, nine phthalates, six parabens, and four BADGEs were determined in indoor dust samples collected from Hanoi, Hatinh, Hungyen, and Thaibinh, in Vietnam. Total concentrations of phthalates, parabens, and BADGEs in indoor dust ranged from 3440 to 106,000 ng/g (median: 22,600 ng/g), 40-840 ng/g (median: 123 ng/g), and 23 to 1750 ng/g (median: 184 ng/g), respectively. Based on the measured median concentration of phthalates, parabens, and BADGEs in indoor dust, we estimated human exposure doses to these compounds through indoor dust ingestion for various age groups. The exposure doses to phthalates, parabens, and BADGEs decreased with age and ranged from 19.4 to 90.4 ng/kg-bw/d, 0.113-0.528 ng/kg-bw/d, and 0.158-0.736 ng/kg-bw/d, respectively. This is the first study on the occurrence and human exposure of phthalates, parabens, and BADGEs in indoor dust from Vietnam.