EMAIL THIS PAGE TO A FRIEND
Biochemical and Biophysical Research Communications
Fisetin induces Sirt1 expression while inhibiting early adipogenesis in 3T3-L1 cells.
PMID 26499075
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F4043
Fisetin, ≥98%
C15H10O6 · xH2O