EMAIL THIS PAGE TO A FRIEND
PeerJ
Autophagic flux inhibition and lysosomogenesis ensuing cellular capture and retention of the cationic drug quinacrine in murine models.
PMID 26500823
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q3251
Quinacrine dihydrochloride, ≥90%
C23H30ClN3O · 2HCl