EMAIL THIS PAGE TO A FRIEND
Diseases of Aquatic Organisms
Histophagous ciliate Pseudocollinia brintoni and bacterial assemblage interaction with krill Nyctiphanes simplex. I. Transmission process.
PMID 26503776