EMAIL THIS PAGE TO A FRIEND
Nanoscale
Destruction of amyloid fibrils by graphene through penetration and extraction of peptides.
PMID 26503908
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
105228
1,1,1,3,3,3-Hexafluoro-2-propanol, ≥99%
C3H2F6O