EMAIL THIS PAGE TO A FRIEND
Journal of Mass Spectrometry : Jms
Simultaneous detection of phosphatidylcholines and glycerolipids using matrix-enhanced surface-assisted laser desorption/ionization-mass spectrometry with sputter-deposited platinum film.
PMID 26505771
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8394
1,2-Dioleoyl-rac-glycerol, ≥97%
C39H72O5
D2135
1,2-Dipalmitoyl-rac-glycerol, ≥99%
C35H68O5