PLoS ONE
Dosage Compensation of X-Linked Muller Element F Genes but Not X-Linked Transgenes in the Australian Sheep Blowfly.
PMID 26506426
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
309435
3-Methyl-1-butanol, anhydrous, ≥99%
C5H12O
320021
3-Methyl-1-butanol, ACS reagent, ≥98.5%
C5H12O
M32658
3-Methyl-1-butanol, reagent grade, 98%
C5H12O
I9392
3-Methylbutanol, BioReagent, for molecular biology, ≥98.5%
C5H12O
59085
3-Methylbutanol, BioUltra, for molecular biology, ≥99.0% (GC)
C5H12O
59090
3-Methylbutanol, puriss. p.a., ACS reagent, ≥98.5% (GC)
C5H12O
C7559
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
CHCl3
372978
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
CHCl3
288306
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
439142
Chloroform, for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
437581
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
CHCl3
480150
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
CHCl3
270636
Chloroform, for HPLC, ≥99.8%
CHCl3
48520-U
Chloroform, analytical standard
CHCl3
05-3433
Chloroform, for residue analysis, JIS 5000
CHCl3
05-3450
Chloroform, for HPLC
CHCl3
W205702
Isoamyl alcohol, ≥98%, FG
C5H12O
W205710
Isoamyl alcohol, natural, ≥98%, FG
C5H12O
P9346
Liquified Phenol, meets USP testing specifications, ≥89.0%
C6H6O
P1037
Phenol, for molecular biology
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
328111
Phenol, unstabilized, purified by redistillation, ≥99%
C6H6O
242322
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
C6H6O
185450
Phenol, unstabilized, ReagentPlus®, ≥99%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
33517
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
C6H6O
16016
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
C6H6O
16017
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
C6H6O
16018
Phenol, ≥96.0% (calc. on dry substance, T)
C6H6O
W322318
Phenol, ≥99%
C6H6O