EMAIL THIS PAGE TO A FRIEND
Journal of dairy science
Milk-clotting mechanism of Dregea sinensis Hemsl. protease.
PMID 26506540
Abstract
Dregea sinensis Hemsl. is used as a milk coagulant to produce goat milk cakes in Yunnan, China. However, the composition of milk-clotting compounds and the related mechanism have not been reported. Crude protease was extracted from the stem, purified, and then separated with a Millipore ultrafiltration centrifuge tube. Cysteine protease (procerain B) was identified as the main milk-clotting protein through electrospray ionization mass spectrometry, and its molecular weight was 23.8 kDa. The protease can partially degrade α-casein (CN) and completely degrade β- and κ-CN, and κ-CN degradation resulted in milk clotting. The molecular weight and AA sequence of the peptide fractions were determined through matrix-assisted laser desorption/ionization time-of-flight mass spectrometry and a peptide sequencer, respectively. The enzyme cleaved κ-CN at Ala90-Gln91 and produced deputy κ-CN and caseinomacropeptide with molecular weights of 12 and 6.9 kDa, respectively. This cleavage site differed from the majority of chymosins cleaved at Phe105-Met106.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
755842
6-Maleimidohexanoic acid, 90% (GC)
C10H13NO4