EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Single- versus Multiple-Pest Infestation Affects Differently the Biochemistry of Tomato (Solanum lycopersicum 'Ailsa Craig').
PMID 26507319