EMAIL THIS PAGE TO A FRIEND
Neurochemical Research
Competition, Selectivity and Efficacy of Analogs of A-84543 for Nicotinic Acetylcholine Receptors with Repositioning of Pyridine Nitrogen.
PMID 26508288