EMAIL THIS PAGE TO A FRIEND
British journal of haematology
Coagulation activation in sickle cell trait: an exploratory study.
PMID 26511074
Abstract
Recent epidemiologic data suggest that sickle cell trait (HbAS; AS) is a risk factor for venous thromboembolism. We conducted an exploratory study of healthy subjects with AS under baseline conditions to determine whether a chronic basal hyperactivation of coagulation exists, and if so, what mechanism(s) contribute to this state. Eighteen healthy AS individuals were compared to 22 African-American controls with a normal haemoglobin profile (HbAA; AA) and 17 patients with sickle cell disease (HbSS; SS). Plasma thrombin-antithrombin complexes and D-dimer levels were elevated in AS relative to AA patients (P = 0·0385 and P = 0·017, respectively), and as expected, were much higher in SSversusAA (P < 0·0001 for both). Thrombin generation in platelet poor plasma was indistinguishable between AA and AS subjects, whereas a paradoxical decrease in endogenous thrombin potential was observed in SS (P ≤ 0·0001). Whole blood tissue factor was elevated in SS compared to AA (P = 0·005), but did not differ between AA and AS. Plasma microparticle tissue factor activity was non-significantly elevated in AS (P = 0·051), but was clearly elevated in SS patients (P = 0·004) when compared to AA controls. Further studies in larger cohorts of subjects with sickle cell trait are needed to confirm the results of this preliminary investigation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3705 Transferrin human, recombinant, expressed in rice