EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
Graphene Functionalized with Arginine Decreases the Development of Glioblastoma Multiforme Tumor in a Gene-Dependent Manner.
PMID 26512645