EMAIL THIS PAGE TO A FRIEND
Chronobiology international
Effects of starvation, re-feeding and timing of food supply on daily rhythm features of gut melatonin in carp (Catla catla).
PMID 26513010
Abstract
Influences of starvation, re-feeding and time of food supply on daily rhythm features of melatonin (5-methoxy-N-acetyltryptamine) and its key regulator AANAT (arylalkylamine N-acetyltransferase) protein in the gut tissues were separately evaluated in carp Catla catla. The first experiment was aimed at demonstration of duration dependent effects of starvation and re-feeding after starvation on the daily profiles and rhythm features of gut melatonin and AANAT. Accordingly, juvenile carp were randomly distributed in three groups, which were (a) provided with balanced diet daily at a fixed time, that is, 10:00 clock hour or zeitgeber time (ZT) 4 (control), or (b) starved (for 2-, 4-, 6- or 8 days), or (c) initially starved for 8 days and then re-fed (for 2-, 4-, 6-, 8-, 12- or 16 days) daily with the same food and at the time (ZT4) used for control fish. The carp in each group were sampled for collection of gut tissues at six different time points at a regular interval of 4 h in a daily cycle. In another experiment, the influences of timing of food supply were separately examined in four fish groups, which were provided with a fixed amount of food once daily either at 06:00 or 12:00 or 18:00 or 24:00 clock hour corresponding to ZT0 or ZT6 or ZT12 or ZT18, respectively, for 7 days before sampling at 12 different time points with a regular interval of 2 h in a 24-h cycle. The study revealed a gradual increase in the mesor and amplitude values of melatonin and AANAT in gut with the progress of starvation till their values reached maximum at day-6 and remained steady thereafter. In contrast, re-feeding of 8-day starved fish resulted in a sharp decrease in their mesor and amplitude values after 2 days and then followed by a steady-state increase till re-attainment of their values close to control fish at the end of 16 days. The acrophase of these gut variables in each control, starved and re-fed fish was noted mostly at midday or ZT6. However, the results of another experiment demonstrated that a shift of food supply time led to a shift in their acrophase. The amount of residual food in the gut lumen in each, but not starved, fish by showing a significant positive correlation independently with the gut levels of melatonin and AANAT also indicated possible role of food as the synchronizer for their daily rhythms. Collectively, it appears reasonable to argue that daily profiles of gut melatonin and AANAT are strongly influenced by the availability of food, while their daily rhythm features seem to be dependent mostly on the time of food supply in carp.