EMAIL THIS PAGE TO A FRIEND
Journal of Experimental & Clinical Cancer Research
Antitumor activity of the multikinase inhibitor regorafenib in patient-derived xenograft models of gastric cancer.
PMID 26514182