EMAIL THIS PAGE TO A FRIEND
Genes & development
Mutation of the TERT promoter, switch to active chromatin, and monoallelic TERT expression in multiple cancers.
PMID 26515115
Abstract
Somatic mutations in the promoter of the gene for telomerase reverse transcriptase (TERT) are the most common noncoding mutations in cancer. They are thought to activate telomerase, contributing to proliferative immortality, but the molecular events driving TERT activation are largely unknown. We observed in multiple cancer cell lines that mutant TERT promoters exhibit the H3K4me2/3 mark of active chromatin and recruit the GABPA/B1 transcription factor, while the wild-type allele retains the H3K27me3 mark of epigenetic silencing; only the mutant promoters are transcriptionally active. These results suggest how a single-base-pair mutation can cause a dramatic epigenetic switch and monoallelic expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48520-U
Chloroform, analytical standard
CHCl3