EMAIL THIS PAGE TO A FRIEND
Journal of cancer research and clinical oncology
MicroRNA-206 functions as a tumor suppressor in colorectal cancer by targeting FMNL2.
PMID 26515696
Abstract
Colorectal cancer (CRC) is one of the most common cancers in the world. MicroRNAs play important roles in the progression of CRC. This study aimed to investigate the role of miR-206 and its novel mechanism in the invasion and metastasis of CRC. Real-time RT-PCR or Western blotting was used to detect the expressions of miR-206, FMNL2 and c-MET in CRC cell lines and tissues. Luciferase reporter assays were conducted to detect the associations between miR-206 and 3'UTRs of FMNL2 and c-MET. A series of loss-of-function and gain-of-function assays were performed to evaluate the effect of miR-206 on the proliferation, invasion and metastasis of CRC cells. miR-206 was significantly down-regulated in CRC tissues and correlated closely with differentiation, lymphatic metastasis and serosal invasion. miR-206 suppressed CRC cell proliferation by arresting CRC cells in the G1/G0 phase and accelerating apoptosis. miR-206 also inhibited cell invasion and lung metastasis in CRC cells. Mechanically, FMNL2 and c-MET were identified as direct targets of miR-206. And FMNL2 rescued the suppression of miR-206 in the proliferation and invasion of CRC cells. This study revealed functional and mechanistic links between miR-206 and oncogene FMNL2 and c-MET in the progression of CRC. miR-206 functioned as a tumor suppressor in the progression of CRC by targeting FMNL2 and c-MET. Restoration of miR-206 expression may represent a promising therapeutic approach for targeting malignant CRC.