EMAIL THIS PAGE TO A FRIEND
Breast Cancer Research and Treatment
Potentiated suppression of Dickkopf-1 in breast cancer by combined administration of the mevalonate pathway inhibitors zoledronic acid and statins.
PMID 26515701
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6892
Farnesyl pyrophosphate ammonium salt, methanol:ammonia solution, ≥95% (TLC)
C15H37N3O7P2