EMAIL THIS PAGE TO A FRIEND
Marine drugs
Bioactive Hydantoin Alkaloids from the Red Sea Marine Sponge Hemimycale arabica.
PMID 26516870
Abstract
In the course of our continuing efforts to identify bioactive secondary metabolites from Red Sea marine invertebrates, we have investigated the sponge Hemimycale arabica. The antimicrobial fraction of an organic extract of the sponge afforded two new hydantoin alkaloids, hemimycalins A and B (2 and 3), together with the previously reported compound (Z)-5-(4-hydroxybenzylidene)imidazolidine-2,4-dione (1). The structures of the compounds were determined by extensive 1D and 2D NMR (COSY, HSQC and HMBC) studies and high-resolution mass spectral determinations. Hemimycalins A (2) and B (3) represent the first examples of the natural N-alkylated hydantoins from the sponge Hemimycale arabica. Compounds 1-3 displayed variable antimicrobial activities against E. coli, S. aureus, and C. albicans. In addition, compound 1 displayed moderate antiproliferative activity against the human cervical carcinoma (HeLa) cell line. These findings provide further insight into the chemical diversity as well as the biological activity of this class of compounds.