EMAIL THIS PAGE TO A FRIEND
International journal of environmental research and public health
Prevalence and Determinants of Metabolic Health in Subjects with Obesity in Chinese Population.
PMID 26516886
Abstract
The study was to investigate the prevalence of metabolic health in subjects with obesity in the Chinese population and to identify the determinants related to metabolic abnormality in obese individuals. 5013 subjects were recruited from seven provincial capitals in China. The obesity and metabolic status were classified based on body mass index (BMI) and the number of abnormalities in common components of metabolic syndrome. 27.9% of individuals with obesity were metabolically healthy. The prevalence of the metabolically healthy obese (MHO) phenotype was significantly decreased with age in women (p trend < 0.001), but not significantly in men (p trend = 0.349). Central obesity (odds ratio [OR] = 4.07, 95% confidence interval [CI] = 1.93-8.59), longer sedentary time (OR = 1.97, 95%CI = 1.27-3.06), and with a family history of obesity related diseases (hypertension, diabetes, dyslipidemia) (OR = 1.85, 95%CI = 1.26-2.71) were significantly associated with having metabolic abnormality in obese individuals. Higher levels of physical activity and more fruit/vegetable intake had decreased ORs of 0.67 (95%CI = 0.45-0.98) and 0.44 (95%CI = 0.28-0.70), respectively. 27.9% of obese participants are in metabolic health. Central obesity, physical activity, sedentary time, fruits/vegetables intake and family history of diseases are the determinants associated with metabolic status in obesity.