EMAIL THIS PAGE TO A FRIEND
ACS Nano
Solid-State Conversion Chemistry of Multicomponent Nanocrystals Cast in a Hollow Silica Nanosphere: Morphology-Controlled Syntheses of Hybrid Nanocrystals.
PMID 26517204