EMAIL THIS PAGE TO A FRIEND
Cancer Medicine
Coculture with astrocytes reduces the radiosensitivity of glioblastoma stem-like cells and identifies additional targets for radiosensitization.
PMID 26518290