EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Rapid screening of haloacetamides in water using salt-assisted liquid-liquid extraction coupled injection-port silylation gas chromatography-mass spectrometry.
PMID 26518495
Abstract
The rapid screening of trace amounts of the nitrogenous disinfection by-products, haloacetamides (HAcAms), in drinking and swimming pool water was performed by a simple and reliable procedure based on salt-assisted liquid-liquid extraction (SALLE) combined with injection-port silylation gas chromatography-mass spectrometry (IPS-GC-MS) method. The optimal SALLE conditions involved the injection of 4-mL of ethyl acetate into a 10-mL water sample (pH 7) containing 3-g of sodium sulfate. After vortex extraction for 1min and centrifugation, 10μL of the extract (mixed with 1μL of MTBSTFA) was directly determined by IPS-GC-MS. The limits of quantitation (LOQs) were determined to be 0.03-0.3μg/L. Precision, as indicated by relative standard deviations (RSDs), was less than 10% for both intra- and inter-day analysis. Accuracy, expressed as the mean extraction recovery, was between 76% and 94%. The SALLE plus IPS-GC-MS was successfully applied to quantitatively determine HAcAms from drinking and swimming pool water samples, and the total concentrations of the compounds ranged from 0.43 to 4.03μg/L.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
176591
Anthracene-d10, 98 atom % D
C14D10
442456
Anthracene-d10, analytical standard
C14D10