EMAIL THIS PAGE TO A FRIEND
Chinese Medical Journal (English Edition)
Caffeine Suppresses Apoptosis of Bladder Cancer RT4 Cells in Response to Ionizing Radiation by Inhibiting Ataxia Telangiectasia Mutated-Chk2-p53 Axis.
PMID 26521794