EMAIL THIS PAGE TO A FRIEND
International journal of biological macromolecules
Anti-tumor and immunomodulatory activity of selenium (Se)-polysaccharide from Se-enriched Grifola frondosa.
PMID 26522247
Abstract
A polysaccharide termed Se-GP11 was extracted and purified from Se-enriched Grifola frondosa in our previous study. This study investigated the characterization, anti-tumor and immunomodulatory activity of Se-GP11. The results showed that Se-GP11 was composed of mannose, glucose and galactose with a molar ratio of 1:4.91:2.41. The weight-average molecular weight (Mw) and weight-average mean square radius (Rw) of Se-GP11 in 0.1M sodium chloride solution were 3.3×10(4)Da and 32.8 nm. Se-GP11 existed as a globular conformation with random coil structure. Se-GP11 had no anti-tumor activity against HepG-2 cells in vitro, and it significantly inhibited the growth of Heps tumor in vivo. Se-GP11 increased the relatively thymus and spleen weights as well as serum necrosis factor-alpha (TNF-α) and interleukin-2 (IL-2) levels. In addition, Se-GP11 promoted the phagocytosis and NO production of RAW264.7 as compared with that of the normal control group. The results revealed that the Se-GP11 may exhibit the anti-tumor through improving immunologic function of the tumor bearing mice.