EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Indirect chiral separation of tryptophan enantiomers by high performance liquid chromatography with indirect chemiluminiscence detection.
PMID 26523665
Abstract
In recent years, the study of chiral compounds in vivo has received much attention. In this study, a novel method based on high performance liquid chromatography (HPLC) coupled with chemiluminescence (CL) detection was developed for the separation of tryptophan (Trp) enantiomers. o-Phthalaldehyde and N-acetyl-l-cysteine were used as chiral derivatization reagents for Trp before it can be detected by HPLC-CL method. The separation was carried out on an ODS column using a mobile phase composed of methanol-0.01mol/L phosphate buffer (40/60, v/v). Under the optimum conditions, satisfactory results were obtained, including complete separation, good relative standard deviations and low detection limits. The applicability of the proposed method has been validated by determining Trp in biological samples. Linear responses (r>0.9990) were observed over the range of 2.5×10(-7) to 1.2×10(-5)g/mL of Trp enantiomers, with quantitation limit of 2.5×10(-7)g/mL. The assay method shows good specificity to Trp enantiomers, and thus it will have great potential application in clinical diagnosis. The mean extraction efficiency of Trp enantiomers in mice plasma samples were 98.48% and 97.40%, respectively. The mean relative standard deviation (RSD) of Trp enantiomers were <3%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
735914
Octylphosphonic acid, 97%
C8H19O3P