EMAIL THIS PAGE TO A FRIEND
Journal of AOAC (Association of Official Analytical Chemists) International
Visual and Real-Time Event-Specific Loop-Mediated Isothermal Amplification Based Detection Assays for Bt Cotton Events MON531 and MON15985.
PMID 26525238