EMAIL THIS PAGE TO A FRIEND
BMC Cancer
Reprogramming energy metabolism and inducing angiogenesis: co-expression of monocarboxylate transporters with VEGF family members in cervical adenocarcinomas.
PMID 26525902