EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
High efficiency, narrow particle size distribution, sub-2 μm based macrocyclic glycopeptide chiral stationary phases in HPLC and SFC.
PMID 26526918
Abstract
State of the art chiral chromatography still employs 3-5 μm bonded or immobilized chiral selectors in 10-25 cm columns. With the availability of 1.9 μm narrow particle size distribution (NPSD) silica, it is now possible to make ever shorter, high efficiency columns practical for sub-minute chiral separations. Three macrocyclic glycopeptides (teicoplanin, teicoplanin aglycone, and vancomycin) were bonded onto 1.9 μm NPSD particles. Such packed columns had ∼80% lower backpressure as compared to polydisperse (PD) 1.7 μm silica materials when using the same mobile phase. The decreased backpressure allowed for diminution of frictional heating and allowed for the use of the 1.9 μm NPSD particle based columns at high flow rates. The 1.9 μm NPSD particle based columns showed up to 190,000 plates m(-1) for chiral molecules and 210,000 plates m(-1) for achiral probes. Representative enantiomeric separations are shown for wide classes of compounds, including different types of amino acids, β-blockers, and pharmaceutically important heterocyclic compounds such as oxazolidinones. Applications in three liquid chromatography modes, namely, reversed phase, polar organic mode and normal phase chiral separations were shown with resolution values ranging from 1.5 to 5.7. Additionally, the same columns were used with supercritical fluid chromatography (SFC) for ultrafast separations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D194255
1,3-Dinitrobenzene, 97% anhydrous basis
C6H4N2O4
442244
1,3-Dinitrobenzene, analytical standard
C6H4N2O4
413364
3-(Triethoxysilyl)propyl isocyanate, 95%
C10H21NO4Si
A8380
5α-Androstan-17β-ol-3-one, ≥97.5%
C19H30O2
45726
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
C2H4O2
W200603
Acetic acid, natural, ≥99.5%, FG
C2H4O2
W200611
Acetic acid, ≥99.5%, FCC, FG
C2H4O2
ARK2183
Acetic acid, glacial, PharmaGrade, USP, JP, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C2H4O2
246654
Heptane, anhydrous, 99%
C7H16
494526
Heptane, biotech. grade, ≥99%
C7H16
32287
Heptane, puriss. p.a., Reag. Ph. Eur., ≥99% n-heptane basis (GC)
C7H16
270512
Heptane, HPLC grade, ≥99%
C7H16
13-0381
Heptane, for HPLC
C7H16
H2198
Heptane, ReagentPlus®, 99%
C7H16
H1758
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
HCl
84429
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
HCl
H9892
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
HCl
2104
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
HCl
W530574
Hydrochloric acid solution, 32 wt. % in H2O, FCC
HCl
295426
Hydrogen chloride, ReagentPlus®, ≥99%
HCl
705551
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
HCl
40104-U Methanolic HCl, 3 M HCl in methanol (3N), for GC derivatization, pkg of 1 × 2 L
HCl
270970
Pyridine, anhydrous, 99.8%
C5H5N
W296600
Pyridine, ≥99%
C5H5N
24-6510
Pyridine, SAJ first grade, ≥99.0%
C5H5N
71699
Sodium hydrosulfite, ≥82% (RT)
Na2O4S2
157953
Sodium hydrosulfite, technical grade
Na2O4S2
244511
Toluene, anhydrous, 99.8%
C7H8
648566
Toluene, for HPLC, ≥99.9%
C7H8
270377
Toluene, HPLC grade, 99.8%
C7H8