EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Identification of Reliable Components in Multivariate Curve Resolution-Alternating Least Squares (MCR-ALS): a Data-Driven Approach across Metabolic Processes.
PMID 26531245
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178837
3-(Trimethylsilyl)-1-propanesulfonic acid sodium salt, 97%
C6H15NaO3SSi