EMAIL THIS PAGE TO A FRIEND
Scientific reports
IFNL3 mRNA structure is remodeled by a functional non-coding polymorphism associated with hepatitis C virus clearance.
PMID 26531896
Abstract
Polymorphisms near the interferon lambda 3 (IFNL3) gene strongly predict clearance of hepatitis C virus (HCV) infection. We analyzed a variant (rs4803217 G/T) located within the IFNL3 mRNA 3' untranslated region (UTR); the G allele (protective allele) is associated with elevated therapeutic HCV clearance. We show that the IFNL3 3' UTR represses mRNA translation and the rs4803217 allele modulates the extent of translational regulation. We analyzed the structures of IFNL3 variant mRNAs at nucleotide resolution by SHAPE-MaP. The rs4803217 G allele mRNA forms well-defined 3' UTR structure while the T allele mRNA is more dynamic. The observed differences between alleles are among the largest possible RNA structural alterations that can be induced by a single nucleotide change and transform the UTR from a single well-defined conformation to one with multiple dynamic interconverting structures. These data illustrate that non-coding genetic variants can have significant functional effects by impacting RNA structure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RDD002
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
C8H18N2O4S
H3375
HEPES, ≥99.5% (titration)
C8H18N2O4S
H4034
HEPES, BioPerformance Certified, ≥99.5% (titration), cell culture tested
C8H18N2O4S
PHG0001
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
RES6003H-B7
HEPES, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production, suitable for cell culture
C8H18N2O4S
H7523
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
C8H18N2O4S
H6147
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
C8H18N2O4S
54457
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
C8H18N2O4S
83264
HEPES buffer solution, 1 M in H2O
C8H18N2O4S
449172
Magnesium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl2Mg
449164
Magnesium chloride, anhydrous, beads, −10 mesh, 99.9% trace metals basis
Cl2Mg
M8266
Magnesium chloride, anhydrous, ≥98%
Cl2Mg
M4880
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Cl2Mg
208337
Magnesium chloride, powder, <200 μm
Cl2Mg
M1028
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Cl2Mg
63069
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Cl2Mg
63036
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Cl2Mg
68475
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Cl2Mg
00457
Magnesium chloride solution, 0.1 M
Cl2Mg
791733
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3KO2
P1190
Potassium acetate, for molecular biology, ≥99.0%
C2H3KO2
P5708
Potassium acetate, BioXtra, ≥99.0%
C2H3KO2
P1222
Potassium acetate, meets USP testing specifications
C2H3KO2
255785
Potassium acetate, 99.98% trace metals basis, ~100 ppm Rb, <50 ppm Cs
C2H3KO2
236497
Potassium acetate, ACS reagent, ≥99.0%
C2H3KO2
P1147
Potassium acetate, ReagentPlus®, ≥99.0%
C2H3KO2
60035
Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT)
C2H3KO2
25059
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
C2H3KO2
W292001
Potassium acetate, 98%
C2H3KO2
95843
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
C2H3KO2