EMAIL THIS PAGE TO A FRIEND
Value in health : the journal of the International Society for Pharmacoeconomics and Outcomes Research
Budget Impact Analysis Of N-Terminal Pro-B-Type Natriuretic Peptide Testing For The Diagnosis And Prognosis Of Heart Failure In Greece.
PMID 26531971
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
469548
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
C8H11F3O2