EMAIL THIS PAGE TO A FRIEND
Value in health : the journal of the International Society for Pharmacoeconomics and Outcomes Research
Endovascular Interventions For Treatment Of Femoropopliteal Peripheral Artery Disease: Updated Budget Impact Analysis For Germany Based On Latest Clincal Evidence.
PMID 26531975
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
469548
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 98%
C8H11F3O2