EMAIL THIS PAGE TO A FRIEND
BMC research notes
Methods of high integrity RNA extraction from cell/agarose construct.
PMID 26537242
Abstract
Agarose hydrogels are widely used for three-dimensional cell scaffolding in tissue engineering and cell biology. Recently, molecular profiles have been obtained with extraction of a minimal volume of RNA using fluorescent-tagged quantitative polymerase chain reaction (qPCR), which requires high integrity RNA. However, the agarose interferes considerably with the quantity and quality of the extracted RNA. Moreover, little is known about RNA integrity when the RNA is extracted from cell/agarose construct. Thus, in order to obtain RNA of sufficient integrity, we examined various extraction methods and addressed reproducible methodologies for RNA extraction from cell/agarose constructs using spectrophotometry and microfluidic capillary electrophoresis. With various extraction methods using a mono-phasic solution of phenol and guanidine isothiocyanate, we evaluated quantity and quality of total RNA from cell/agarose construct. Extraction with solution of phenol and guanidine isothiocyanate followed by a silica based membrane filter column gave sufficient RNA integrity number, which allowed us to proceed to fluorescent-tagged qPCR for evaluating various cellular activities. The RNA extraction methods using phenol and guanidine isothiocyanate solution and a silica membrane column can be useful for obtaining high integrity RNA from cell/agarose constructs rich in polysaccharide and extracellular matrix. Our study contributes to further investigation using agarose hydrogels and other materials rich in polysaccharide in the field of cellular and tissue engineering.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0125
Cacodylic acid, ≥98%
C2H7AsO2
20835
Cacodylic acid, ≥99.0%
C2H7AsO2
05-0015
Cacodylic acid, SAJ first grade
C2H7AsO2
P9346
Liquified Phenol, meets USP testing specifications, ≥89.0%
C6H6O
T1406
o-Toluidine, liquid
C7H9N
185426
o-Toluidine, ≥99%
C7H9N
466190
o-Toluidine, 98%
C7H9N
P1037
Phenol, for molecular biology
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
328111
Phenol, unstabilized, purified by redistillation, ≥99%
C6H6O
242322
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
C6H6O
185450
Phenol, unstabilized, ReagentPlus®, ≥99%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
33517
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
C6H6O
16016
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
C6H6O
16017
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
C6H6O
16018
Phenol, ≥96.0% (calc. on dry substance, T)
C6H6O
W322318
Phenol, ≥99%
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
P4557
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
C6H6O
P4682
Phenol solution, BioReagent, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
C6H6O
48688
Phenol solution, certified reference material, 500 μg/mL in methanol
C6H6O
40063
Phenol solution, 5000 μg/mL in methanol, certified reference material
C6H6O
P2713
Potassium thiocyanate, BioXtra, ≥99.0%
CKNS
207799
Potassium thiocyanate, ACS reagent, ≥99.0%
CKNS
P3011
Potassium thiocyanate, ReagentPlus®, ≥99.0%
CKNS
31272
Potassium thiocyanate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
CKNS