EMAIL THIS PAGE TO A FRIEND
Journal of Translational Medicine
Application of Nanotrap technology for high sensitivity measurement of urinary outer surface protein A carboxyl-terminus domain in early stage Lyme borreliosis.
PMID 26537892