EMAIL THIS PAGE TO A FRIEND
The Journal of Experimental Biology
Chemical cues from fish heighten visual sensitivity in larval crabs through changes in photoreceptor structure and function.
PMID 26538174