EMAIL THIS PAGE TO A FRIEND
Scientific reports
Molecular changes in the medial prefrontal cortex and nucleus accumbens are associated with blocking the behavioral sensitization to cocaine.
PMID 26538265
Abstract
Previous studies have demonstrated that cocaine-induced behavioral sensitization is associated with persistent functional and structural alterations in the medial prefrontal cortex (mPFC) and nucleus accumbens (NAc); however, the molecular mechanisms underlying these changes have not been elucidated. In this study, the behavioral sensitization to cocaine was established in Sprague Dawley rats and was measured by locomotion and behavioral rating. The brain tissue homogenization was used for measuring the level of brain-derived neurotrophic factor (BDNF), the expression and activity of integrin-linked kinase (ILK), level of protein kinase B (Akt) phosphorylation at serine 473 and threonine 308, and the expression of p75(NTR), TrkA, and TrkB protein. The Results showed that cocaine sensitization was associated with increased BDNF, ILK activity, phospho-Akt Ser(473), p75(NTR), and TrkB protein levels in the mPFC and NAc core. The combination of pergolide and ondansetron normalized not only behavioral sensitization, but also the increases in these molecular markers. Dual immunofluoresence staining showed that ILK expression is co-distributed with p75(NTR) and TrkA expression in both the mPFC and NAc core. Results suggested that the BDNF-TrkA/p75(NTR)-ILK-Akt signaling pathway may be active in cocaine sensitization and associated neural plasticity in the mPFC and NAc core.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3639
Ondansetron hydrochloride dihydrate, ≥98% (HPLC), powder
C18H19N3O · HCl · 2H2O