EMAIL THIS PAGE TO A FRIEND
Journal of environmental health science & engineering
Optimization of sonochemical degradation of tetracycline in aqueous solution using sono-activated persulfate process.
PMID 26539297