EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Sciences
Genotoxicity of Superparamagnetic Iron Oxide Nanoparticles in Granulosa Cells.
PMID 26540051
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
700312
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
700320
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
700304
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Fe3O4
725331
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
725358
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
725366
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Fe3O4
747327
Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747424
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm diameter, biotin functionalized, 1 mg/mL Fe, dispersion in H2O
Fe3O4
747254
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), carboxylic acid functionalized, 5 mg/mL Fe in H2O, dispersion
Fe3O4
747343
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747319
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747300
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size (TEM), amine functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
747408
Iron oxide(II,III), magnetic nanoparticles solution, 30 nm avg. part. size (TEM), PEG functionalized, dispersion, 1 mg/mL Fe in H2O
Fe3O4
747416
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), biotin functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
790508
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size (TEM), PEG functionalized, 1 mg/mL Fe in H2O, dispersion
Fe3O4
518158
Iron(II,III) oxide, 99.99% trace metals basis
Fe3O4
637106
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Fe3O4
310069
Iron(II,III) oxide, powder, <5 μm, 95%
Fe3O4