EMAIL THIS PAGE TO A FRIEND
Scientific reports
Rapid and robust generation of long-term self-renewing human neural stem cells with the ability to generate mature astroglia.
PMID 26541394
Abstract
Induced pluripotent stem cell bear the potential to differentiate into any desired cell type and hold large promise for disease-in-a-dish cell-modeling approaches. With the latest advances in the field of reprogramming technology, the generation of patient-specific cells has become a standard technology. However, directed and homogenous differentiation of human pluripotent stem cells into desired specific cell types remains an experimental challenge. Here, we report the development of a novel hiPSCs-based protocol enabling the generation of expandable homogenous human neural stem cells (hNSCs) that can be maintained under self-renewing conditions over high passage numbers. Our newly generated hNSCs retained differentiation potential as evidenced by the reliable generation of mature astrocytes that display typical properties as glutamate up-take and expression of aquaporin-4. The hNSC-derived astrocytes showed high activity of pyruvate carboxylase as assessed by stable isotope assisted metabolic profiling. Moreover, using a cell transplantation approach, we showed that grafted hNSCs were not only able to survive but also to differentiate into astroglial in vivo. Engraftments of pluripotent stem cells derived from somatic cells carry an inherent tumor formation potential. Our results demonstrate that hNSCs with self-renewing and differentiation potential may provide a safer alternative strategy, with promising applications especially for neurodegenerative disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5499
Dorsomorphin, ≥98% (HPLC)
C24H25N5O
A5960
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
C6H8O6
A7506
L-Ascorbic acid, reagent grade, crystalline
C6H8O6
A0278
L-Ascorbic acid, reagent grade
C6H8O6
A4403
L-Ascorbic acid, powder, cell culture tested, γ-irradiated
C6H8O6
A4544
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
C6H8O6
A2218
L-Ascorbic acid, meets USP testing specifications
C6H8O6
A2174
L-Ascorbic acid, plant cell culture tested
C6H8O6
95209
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
C6H8O6
33034
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.7-100.5% (oxidimetric)
C6H8O6
95210
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
C6H8O6
47863
L-Ascorbic acid, analytical standard
C6H8O6
255564
L-Ascorbic acid, ACS reagent, ≥99%
C6H8O6
A92902
L-Ascorbic acid, 99%
C6H8O6
W210901
L-Ascorbic acid, FCC, FG
C6H8O6
G5667
L-Glutamic acid, PharmaGrade, Ajinomoto, EP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C5H9NO4
W328502
L-Glutamic acid, 99%, FCC
C5H9NO4
G8415
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
C5H9NO4
49449
L-Glutamic acid, BioUltra, ≥99.5% (NT)
C5H9NO4
G1251
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
C5H9NO4
P3766
Phorbol 12-myristate 13-acetate 4-O-methyl ether
C37H58O8
V-011
Retinol solution, 100 μg/mL ± 25% (Refer to COA) (Ethanol with 0.1% (w/v) BHT), ampule of 1 mL, certified reference material
C20H30O