PloS one
Identification of Small-Molecule Inhibitors against Meso-2, 6-Diaminopimelate Dehydrogenase from Porphyromonas gingivalis.
PMID 26544875
Abstract
Species-specific antimicrobial therapy has the potential to combat the increasing threat of antibiotic resistance and alteration of the human microbiome. We therefore set out to demonstrate the beginning of a pathogen-selective drug discovery method using the periodontal pathogen Porphyromonas gingivalis as a model. Through our knowledge of metabolic networks and essential genes we identified a "druggable" essential target, meso-diaminopimelate dehydrogenase, which is found in a limited number of species. We adopted a high-throughput virtual screen method on the ZINC chemical library to select a group of potential small-molecule inhibitors. Meso-diaminopimelate dehydrogenase from P. gingivalis was first expressed and purified in Escherichia coli then characterized for enzymatic inhibitor screening studies. Several inhibitors with similar structural scaffolds containing a sulfonamide core and aromatic substituents showed dose-dependent inhibition. These compounds were further assayed showing reasonable whole-cell activity and the inhibition mechanism was determined. We conclude that the establishment of this target and screening strategy provides a model for the future development of new antimicrobials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74875
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
C24H21Na3O11S3
B0126
Bromophenol Blue
C19H10Br4O5S
114391
Bromophenol Blue, ACS reagent
C19H10Br4O5S
E5389
Erythromycin, BioReagent, suitable for cell culture
C37H67NO13
E6376
Erythromycin, potency: ≥850 μg per mg
C37H67NO13
E0774
Erythromycin, meets USP testing specifications
C37H67NO13
45703
Erythromycin standard solution, 1 mg/mL in H2O
C37H67NO13
G2289
Glycerin, meets USP testing specifications
C3H8O3
G5516
Glycerol, for molecular biology, ≥99.0%
C3H8O3
G9012
Glycerol, ≥99.5%
C3H8O3
G2025
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
C3H8O3
G6279
Glycerol, BioXtra, ≥99% (GC)
C3H8O3
49767
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
C3H8O3
W252506
Glycerol, FCC, FG
C3H8O3
49781
Glycerol solution, 86-89% (T)
C3H8O3 · xH2O
I2399
Imidazole, ACS reagent, ≥99% (titration)
C3H4N2
I5513
Imidazole, for molecular biology, ≥99% (titration)
C3H4N2
56750
Imidazole, puriss. p.a., ≥99.5% (GC)
C3H4N2
I202
Imidazole, ReagentPlus®, 99%
C3H4N2
56749
Imidazole, BioUltra, ≥99.5% (GC)
C3H4N2
792527
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
C3H4N2
56748
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
C3H4N2
I0250
Imidazole, ≥99% (titration), crystalline
C3H4N2
68268
Imidazole buffer Solution, BioUltra, 1 M in H2O
C3H4N2
T2501
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
C15H10I4NNaO4 · 5H2O
T0397
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
C15H10I4NNaO4 · 5H2O
658685
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
OsO4
201030
Osmium tetroxide, ReagentPlus®, 99.8%
O4Os
O5500
Osmium tetroxide, Sealed ampule.
O4Os
419494
Osmium tetroxide, ACS reagent, ≥98.0%
O4Os