EMAIL THIS PAGE TO A FRIEND
PLoS Computational Biology
A Kinetic Platform to Determine the Fate of Hydrogen Peroxide in Escherichia coli.
PMID 26545295
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8285
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
C21H27N7O14P2
N8410
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
C21H27N7O14P2
N8535
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
C21H27N7O14P2
N7004
β-Nicotinamide adenine dinucleotide hydrate, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
C21H27N7O14P2 · xH2O
N6522
β-Nicotinamide adenine dinucleotide hydrate, ≥98%, BioUltra, from yeast
C21H27N7O14P2 · xH2O
N1636
β-Nicotinamide adenine dinucleotide hydrate, purified by column chromatography, ≥99%
C21H27N7O14P2 · xH2O
N3014
β-Nicotinamide adenine dinucleotide hydrate, cell culture tested, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
C21H27N7O14P2 · xH2O
N7381
β-Nicotinamide adenine dinucleotide hydrate, Grade AA-1, ≥95% (HPLC)
C21H27N7O14P2 · xH2O
N1511
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
C21H27N7O14P2 · xH2O
43410
β-Nicotinamide adenine dinucleotide hydrate, ≥95% (HPLC)
C21H27N7O14P2 · xH2O
N7132
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%
C21H26LiN7O14P2
316989
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
H2O2
H1009
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
H2O2
40287
Hydrogen peroxide solution, semiconductor grade MOS PURANAL (Honeywell 17937), ≥30%
H2O2
18304
Hydrogen peroxide solution, meets analytical specification of Ph. Nord., 34.5-36.5%
H2O2