EMAIL THIS PAGE TO A FRIEND
The Science of the Total Environment
Comparing a silver-impregnated activated carbon with an unmodified activated carbon for disinfection by-product minimisation and precursor removal.
PMID 26546763