EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Multimodal super-resolution optical microscopy visualizes the close connection between membrane and the cytoskeleton in liver sinusoidal endothelial cell fenestrations.
PMID 26549018