EMAIL THIS PAGE TO A FRIEND
Scientific Reports
Plasma biomarker for detection of early stage pancreatic cancer and risk factors for pancreatic malignancy using antibodies for apolipoprotein-AII isoforms.
PMID 26549697
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I9516
2-Propanol, BioReagent, for molecular biology, ≥99.5%
C3H8O
278475
2-Propanol, anhydrous, 99.5%
C3H8O
675431
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
C3H8O
437522
2-Propanol, ACS reagent, ≥99.5%
C3H8O
59304
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
C3H8O
40301
2-Propanol, semiconductor grade VLSI PURANAL (Honeywell 17604)
C3H8O
40219
2-Propanol, semiconductor grade PURANAL (Honeywell 17829)
C3H8O
270490
2-Propanol, 99.5%, HPLC grade
C3H8O
15-2360
2-Propanol, for HPLC
C3H8O
733458
2-Propanol, electronic grade, 99.999% trace metals basis
C3H8O
270717
Acetonitrile, HPLC grade, ≥99.93%
C2H3N
L010000-HH
Acetonitrile, for DNA synthesis
C2H3N
01-0650
Acetonitrile, ≥99.8%, for HPLC
C2H3N
01-0623
Acetonitrile, for residue analysis, JIS 5000
C2H3N
01-0645
Acetonitrile, for chromatography
C2H3N
733466
Acetonitrile, electronic grade, 99.999% trace metals basis
C2H3N
L110001
Acetonitrile
C2H3N
L011800
Acetonitrile
C2H3N
639133
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
C2H3N
576956
Acetonitrile solution, contains 0.1 % (v/v) formic acid, for HPLC
C2H3N
574732
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, for HPLC
C2H3N
574724
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
C2H3N
679186
Acetonitrile solution, contains 5 % (v/v) water, 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, for HPLC
C2H3N
685461
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
C2H3N
703664
Acetonitrile solution, NMR reference standard, 0.23 wt. % in D2O (99.9 atom % D), water 0.05 wt. %, NMR tube size 6.5 mm × 8 in.
C2H3N
2107
E-Toxate Water, endotoxin, free
H2O
W292907
Isopropyl alcohol, ≥99.7%, FCC, FG
C3H8O
30-4310
Trifluoroacetic acid, SAJ special grade, ≥99.0%
C2HF3O2
91709
Trifluoroacetic acid solution, BioReagent, suitable for protein sequencing, ~25% in H2O
C2HO2F3
W4502
Water, Molecular Biology Reagent
H2O