EMAIL THIS PAGE TO A FRIEND
Journal of inclusion phenomena and macrocyclic chemistry
Synthesis of thiol derivatives of azobenzocrown ethers. The preliminary studies on recognition of alkali metal ions by gold nanoparticles functionalized with azobenzocrown and lipoic acid.
PMID 26549980
Abstract
The article presents the synthesis of novel 13- and 16-membered azobenzocrown derivatives with peripheral thiol moieties and preliminary studies assessing their possible application in plasmonic sensors based on gold nanoparticles. The effect of the length of the chain connecting the macrocycle with the thiol group and the effect of the presence of the additional functional compound, i.e. lipoic acid, on the sensor response was analyzed. Colloidal gold nanoparticles modified with a 16-membered crown with a thiol group on oxyethylene (compound