EMAIL THIS PAGE TO A FRIEND
Journal of ophthalmology
A Comparative Study between Vitrectomy with Internal Tamponade and a New Modified Fiber Optic Illuminated Ando Plombe for Cases of Macular Hole Retinal Detachment in Myopic Eyes.
PMID 26550487
Abstract
Aim. To compare pars plana vitrectomy (PPV) with silicone tamponade or gas (Groups Ia and Ib) and a new modified Ando plombe equipped with a fiber optic light (Group II) for cases with macular hole retinal detachment (MHRD) in high myopic eyes (axial length > 26 mm). Methods. A prospective interventional randomized case series included 60 eyes (20 in each group). Successful outcome was considered if the retina was completely attached at the end of the follow-up period. Complications were identified for each group. Results. Visual acuity improved by 37.31%, 40.67%, and 49.40% in Groups Ia, Ib, and II, respectively. The success rate was 55%, 60%, and 100% in Groups Ia , Ib, and II, respectively, with a statistically significant difference between Groups Ia, Ib, and II (p < 0.001 in Ia, p: 0.002 in Ib). Complications rates were 60%, 45%, and 20% in Groups Ia, Ib, and II, respectively, with a statistically significant difference between Groups Ia and II (p: 0.01). Conclusion. Fiber optic illuminated Ando plombe allows better positioning under the macula and consequently improves the success rate of epimacular buckling in comparison to PPV with internal tamponade in MMHRD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295701
Sulfur hexafluoride, ≥99.75%
F6S