EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Capillary electrophoresis with end-column electrochemiluminescence for ultrasensitive determination of urapidil hydrochloride in rat plasma and its application to pharmacokinetics study.
PMID 26551206
Abstract
A simple, sensitive and selective method for determination of urapidil hydrochloride was developed using capillary electrophoresis with electrochemiluminescence (CE-ECL) technique for the first time. Under the optimized experimental conditions, the ECL intensity was linear with the concentration of urapidil hydrochloride in the range from 0.050 to 50.0ng/mL and the detection limit was 0.014ng/mL (S/N=3). The proposed method was used for studying pharmacokinetics of urapidil hydrochloride in rat plasma and the main pharmacokinetic parameters of the peak concentration (Cmax), half life time (T1/2) and peak concentration time (Tmax) were 240.45±21.15ng/mL, 0.58±0.16h and 1.08±0.13h, respectively. The recoveries of urapidil hydrochloride in the diluted extracts of rat plasma samples ranged from 96.68 to 98.82%. The RSD was lower than 3%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D216305
2,2′-Bipyridyl, ReagentPlus®, ≥99%
C10H8N2
U100
Urapidil hydrochloride, solid
C20H29N5O3 · HCl