EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Calcium-sensing receptor (CaSR)-mediated anti-inflammatory effects of L-amino acids in intestinal epithelial cells.
PMID 26551350
Abstract
Calcium-sensing receptor (CaSR) plays an essential role in sensing nutrients and monitoring ion balance in the human gut. However, no discovery of CaSR-mediated anti-inflammatory effect of l-amino acids (l-AAs) on the gut system has been reported. The aim of this study is to screen and identify the anti-inflammatory activity of various l-AAs in intestinal epithelial cells (IECs) and stepwise illustrate a possible molecular mechanism for anti-inflammation. We used Caco-2 and HT-29 cell lines to evaluate the anti-inflammatory activity of l-AAs and revealed that l-tryptophan (l-Trp) and l-valine (l-Val) have strong anti-inflammatory activity consistent in both cell lines. l-Trp treatment (5 mM) reduced TNF-α-induced IL-8 secretion from HT-29 or Caco-2 cells to about 50 or 40%, respectively. l-Trp also significantly inhibited the expression of phosphorylation of JNK or IκBα to around 50% in HT-29 cells. However, the above inhibitory effects of l-Trp on inflammatory responses in TNF-α-induced HT-29 cells were abrogated by NPS-2143. The result of CaSR antagonist NPS-2143 pretreatment study suggests l-Trp exerts anti-inflammatory effects on IECs through CaSR activation. The involvement of β-arrestin2 was then found to block tumor necrosis factor (TNF)-α-induced signaling pathways after CaSR activated by l-Trp. These results validate a novel mechanism underlying CaSR agonistic l-AAs exerting anti-inflammatory effects on human intestinal epithelia.