EMAIL THIS PAGE TO A FRIEND
Radiation Oncology (London, England)
Combining carbon ion irradiation and non-homologous end-joining repair inhibitor NU7026 efficiently kills cancer cells.
PMID 26553138
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1537
NU7026, ≥98% (HPLC), solid
C17H15NO3