EMAIL THIS PAGE TO A FRIEND
Microbial cell factories
Molecular genetics of naringenin biosynthesis, a typical plant secondary metabolite produced by Streptomyces clavuligerus.
PMID 26553209
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5893
(±)-Naringenin, ≥95%
C15H12O5
W530098
Naringenin, natural (US), 98%
C15H12O5